Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113359.4 | NP_566749.1 | 2 - 3 | 514 | 99 | |
GenBank mRNA | AY056398.1 | - | 2 - 3 | 285 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |