Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113392.3 | NP_189124.1 | 3 - 4 | 440 | 108 | |
GenBank mRNA | AJ224307.1 | - | 3 - 4 | 417 | 108 | |
AY080799.1 | - | 3 - 4 | 438 | 108 | ||
AY114077.1 | - | 3 - 4 | 296 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |