Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203042.1 | NP_001189971.1 | 10 - 11 | 1881 | 131 | |
NM_113453.3 | NP_189183.2 | 10 - 11 | 1927 | 131 | ||
NM_202631.2 | NP_974360.1 | 10 - 11 | 1712 | 131 | ||
GenBank mRNA | AY075617.1 | - | 10 - 11 | 1660 | 131 | |
BT002619.1 | - | 10 - 11 | 1581 | 131 | ||
FJ708728.1 | - | 10 - 11 | 1581 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |