Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203671.1 | NP_001190600.1 | 12 - 13 | 1449 | 83 | |
NM_125674.3 | NP_201085.1 | 11 - 12 | 1434 | 83 | ||
GenBank mRNA | AF148852.1 | - | 11 - 12 | 1342 | 83 | |
AY045634.1 | - | 11 - 12 | 1381 | 83 | ||
AY050802.1 | - | 11 - 12 | 1380 | 83 | ||
AY054682.1 | - | 11 - 12 | 1342 | 83 | ||
AY081453.1 | - | 11 - 12 | 1257 | 83 | ||
AY084775.1 | - | 11 - 12 | 1342 | 83 | ||
AY091405.1 | - | 11 - 12 | 1257 | 83 | ||
AY098952.1 | - | 11 - 12 | 1257 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |