Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP94B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345592.1 | NP_001331365.1 | 1 | 1703 | 102 | |
NM_125740.3 | NP_201150.1 | 1 | 1647 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |