Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125752.4 | NP_201162.1 | 1 - 2 | 333 | 91 | |
GenBank mRNA | AY102109.1 | - | 1 - 2 | 256 | 91 | |
AY139804.1 | - | 1 - 2 | 176 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |