Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203680.1 | NP_001190609.1 | 1 - 2 | 178 | 85 | |
NM_125762.4 | NP_568976.1 | 1 - 2 | 159 | 85 | ||
GenBank mRNA | AY035161.1 | - | 1 - 2 | 124 | 85 | |
AY063032.1 | - | 1 - 2 | 93 | 85 | ||
AY084523.1 | - | 1 - 2 | 122 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |