Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203680.1 | NP_001190609.1 | 2 - 3 | 264 | 68 | |
NM_125762.4 | NP_568976.1 | 2 - 3 | 245 | 68 | ||
GenBank mRNA | AY035161.1 | - | 2 - 3 | 210 | 68 | |
AY063032.1 | - | 2 - 3 | 179 | 68 | ||
AY084523.1 | - | 2 - 3 | 208 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |