Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI-4KBETA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345618.1 | NP_001330874.1 | 4 - 5 | 2474 | 74 | |
NM_125803.4 | NP_201212.1 | 4 - 5 | 2506 | 74 | ||
GenBank mRNA | AF462854.1 | - | 4 - 5 | 2474 | 74 | |
AY139795.1 | - | 4 - 5 | 1930 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |