Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125857.3 | NP_201266.1 | 3 - 4 | 625 | 68 | |
GenBank mRNA | AB493812.1 | - | 3 - 4 | 459 | 68 | |
AY099684.1 | - | 3 - 4 | 486 | 68 | ||
BT000277.1 | - | 3 - 4 | 459 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |