Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125871.5 | NP_201280.1 | 1 - 2 | 672 | 63 | |
GenBank mRNA | AY140092.1 | - | 1 - 2 | 582 | 63 | |
BT008480.1 | - | 1 - 2 | 442 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |