Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125877.4 | NP_568995.2 | 2 - 3 | 550 | 77 | |
GenBank mRNA | AB493813.1 | - | 2 - 3 | 382 | 77 | |
AF426252.1 | - | 2 - 3 | 382 | 77 | ||
BT025755.1 | - | 2 - 3 | 382 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |