Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125882.3 | NP_201289.1 | 3 - 4 | 1870 | 107 | |
GenBank mRNA | AK227200.1 | - | 3 - 4 | 1392 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |