Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126027.1 | NP_001119499.1 | 1 - 2 | 238 | 233 | |
NM_001345664.1 | NP_001332255.1 | 1 - 2 | 304 | 140 | ||
NM_001345665.1 | NP_001332256.1 | 1 - 2 | 238 | 233 | ||
NM_125906.4 | NP_201312.2 | 1 - 2 | 305 | 140 | ||
GenBank mRNA | AY231450.1 | - | 1 - 2 | 238 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |