Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125910.6 | NP_201316.1 | 6 - 7 | 2126 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |