Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSAC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125999.3 | NP_201403.2 | 3 - 4 | 383 | 90 | |
GenBank mRNA | AF266458.1 | - | 3 - 4 | 349 | 90 | |
AY227249.1 | - | 3 - 4 | 349 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |