Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123796.1 | NP_001117268.1 | 9 - 10 | 1293 | 115 | |
NM_001331977.1 | NP_001320358.1 | 8 - 9 | 991 | 115 | ||
NM_101004.4 | NP_172598.1 | 9 - 10 | 1341 | 115 | ||
GenBank mRNA | AF369563.1 | - | 9 - 10 | 1041 | 115 | |
AK226449.1 | - | 9 - 10 | 1294 | 115 | ||
AY086586.1 | - | 9 - 10 | 1252 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |