Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332010.1 | NP_001322792.1 | 2 - 3 | 1147 | 69 | |
NM_101062.4 | NP_172655.1 | 2 - 3 | 519 | 69 | ||
GenBank mRNA | AY056387.1 | - | 2 - 3 | 394 | 69 | |
AY056403.1 | - | 2 - 3 | 394 | 69 | ||
AY063974.1 | - | 2 - 3 | 394 | 69 | ||
AY088657.1 | - | 2 - 3 | 394 | 69 | ||
BT001980.1 | - | 1 - 2 | 345 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |