Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332035.1 | NP_001321345.1 | 2 - 3 | 255 | 179 | |
NM_101114.4 | NP_172704.1 | 3 - 4 | 512 | 179 | ||
GenBank mRNA | AF528064.1 | - | 3 - 4 | 256 | 179 | |
AK228687.1 | - | 3 - 4 | 429 | 179 | ||
BT026518.1 | - | 3 - 4 | 218 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |