Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035952.3 | NP_001031029.1 | 4 - 5 | 801 | 101 | |
NM_001035953.2 | NP_001031030.1 | 4 - 5 | 808 | 101 | ||
NM_101123.4 | NP_563910.2 | 5 - 6 | 779 | 101 | ||
GenBank mRNA | AK175663.1 | - | 4 - 5 | 795 | 101 | |
AK176660.1 | - | 4 - 5 | 782 | 101 | ||
AK221656.1 | - | 5 - 6 | 699 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |