Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332096.1 | NP_001318998.1 | 4 - 5 | 652 | 83 | |
NM_101255.2 | NP_172842.1 | 3 - 4 | 721 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |