Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198062.1 | NP_001184991.1 | 2 - 3 | 1022 | 84 | |
NM_101264.4 | NP_172851.1 | 2 - 3 | 966 | 84 | ||
GenBank mRNA | U56140.1 | - | 2 - 3 | 957 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |