Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101266.4 | NP_172853.1 | 4 - 5 | 858 | 95 | |
GenBank mRNA | BT026491.1 | - | 4 - 5 | 610 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |