Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101303.4 | NP_563949.1 | 6 - 7 | 721 | 86 | |
GenBank mRNA | AF332450.1 | - | 6 - 7 | 583 | 86 | |
AY059942.1 | - | 6 - 7 | 706 | 86 | ||
AY114599.1 | - | 6 - 7 | 583 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |