Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160865.1 | NP_001154337.1 | 2 - 3 | 198 | 65 | |
NM_001332122.1 | NP_001320938.1 | 2 - 3 | 508 | 65 | ||
NM_001332123.1 | NP_001320937.1 | 2 - 3 | 508 | 65 | ||
NM_101319.4 | NP_172904.2 | 3 - 4 | 408 | 65 | ||
GenBank mRNA | AK175115.1 | - | 2 - 3 | 270 | 65 | |
AK176690.1 | - | 2 - 3 | 270 | 65 | ||
AY136388.1 | - | 2 - 3 | 211 | 65 | ||
BT000187.1 | - | 2 - 3 | 151 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |