Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECOY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160866.1 | NP_001154338.1 | 3 - 4 | 397 | 127 | |
NM_101329.4 | NP_172914.1 | 3 - 4 | 501 | 127 | ||
GenBank mRNA | AY127001.1 | - | 3 - 4 | 397 | 127 | |
BT001031.1 | - | 3 - 4 | 294 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |