Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101352.3 | NP_172936.1 | 6 - 7 | 798 | 139 | |
GenBank mRNA | AJ304841.1 | - | 6 - 7 | 722 | 139 | |
AY039955.1 | - | 6 - 7 | 683 | 139 | ||
AY150494.1 | - | 6 - 7 | 522 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |