Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP70-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198067.2 | NP_001184996.1 | 1 - 2 | 379 | 108 | |
NM_101353.2 | NP_563959.1 | 1 - 2 | 396 | 108 | ||
GenBank mRNA | AM086441.1 | - | 1 - 2 | 310 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |