Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106103.4 | NP_177583.1 | 1 - 2 | 377 | 125 | |
GenBank mRNA | AF217205.2 | - | 1 - 2 | 136 | 125 | |
AK118381.1 | - | 1 - 2 | 367 | 125 | ||
AY519570.1 | - | 1 - 2 | 136 | 125 | ||
BT005272.1 | - | 1 - 2 | 136 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |