Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NdhO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106146.5 | NP_565093.1 | 3 - 4 | 400 | 100 | |
GenBank mRNA | AF344320.1 | - | 3 - 4 | 343 | 100 | |
AF410289.1 | - | 3 - 4 | 400 | 100 | ||
AY143808.1 | - | 3 - 4 | 343 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |