Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH10A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198470.1 | NP_001185399.1 | 12 - 13 | 1410 | 90 | |
NM_106150.4 | NP_565094.1 | 12 - 13 | 1518 | 90 | ||
GenBank mRNA | AY087395.1 | - | 12 - 13 | 1425 | 90 | |
AY093071.1 | - | 12 - 13 | 1383 | 90 | ||
BT008872.1 | - | 12 - 13 | 1254 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |