Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106159.3 | NP_565098.1 | 3 - 4 | 996 | 75 | |
NM_202415.2 | NP_974144.1 | 2 - 3 | 1720 | 75 | ||
GenBank mRNA | AY084709.1 | - | 3 - 4 | 822 | 75 | |
AY099547.1 | - | 3 - 4 | 822 | 75 | ||
BT001236.1 | - | 2 - 3 | 693 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |