Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDSP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106257.3 | NP_177735.1 | 1 - 2 | 740 | 122 | |
GenBank mRNA | AJ318055.1 | - | 1 - 2 | 521 | 122 | |
BT024860.1 | - | 1 - 2 | 502 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |