Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334779.1 | NP_001320598.1 | 4 - 5 | 743 | 71 | |
NM_001334781.1 | NP_001320596.1 | 2 - 3 | 239 | 71 | ||
NM_106391.2 | NP_177866.2 | 5 - 6 | 650 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |