Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334818.1 | NP_001322316.1 | 4 - 5 | 661 | 64 | |
NM_106458.4 | NP_177932.1 | 4 - 5 | 747 | 64 | ||
GenBank mRNA | BT002801.1 | - | 4 - 5 | 656 | 64 | |
BT004348.1 | - | 4 - 5 | 595 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |