Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334832.1 | NP_001319403.1 | 9 - 10 | 3348 | 84 | |
NM_001334833.1 | NP_001322931.1 | 8 - 9 | 3307 | 84 | ||
NM_001334834.1 | NP_001322932.1 | 8 - 9 | 3274 | 84 | ||
NM_106505.5 | NP_177979.1 | 8 - 9 | 1538 | 84 | ||
GenBank mRNA | KJ138926.1 | - | 7 - 8 | 1332 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |