Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334878.1 | NP_001321640.1 | 4 - 5 | 1272 | 103 | |
NM_001334879.1 | NP_001321639.1 | 4 - 5 | 1267 | 103 | ||
NM_001334880.1 | NP_001321641.1 | 4 - 5 | 630 | 103 | ||
NM_106577.4 | NP_565205.1 | 4 - 5 | 983 | 103 | ||
GenBank mRNA | AF326910.1 | - | 4 - 5 | 577 | 103 | |
AY113874.1 | - | 4 - 5 | 296 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |