Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198506.1 | NP_001185435.1 | 47 - 48 | 5155 | 61 | |
NM_001198507.1 | NP_001185436.1 | 48 - 49 | 5218 | 61 | ||
NM_106578.3 | NP_178048.2 | 47 - 48 | 5114 | 61 | ||
GenBank mRNA | EF426860.1 | - | 47 - 48 | 4945 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |