Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124150.1 | NP_001117622.1 | 16 - 17 | 2770 | 72 | |
NM_001198512.1 | NP_001185441.1 | 16 - 17 | 2943 | 72 | ||
NM_001198513.2 | NP_001185442.1 | 16 - 17 | 2883 | 72 | ||
NM_001334925.1 | NP_001321438.1 | 16 - 17 | 2889 | 72 | ||
NM_106632.4 | NP_178101.3 | 16 - 17 | 2870 | 72 | ||
GenBank mRNA | AK226727.1 | - | 16 - 17 | 2772 | 72 | |
EU249329.1 | - | 15 - 16 | 2613 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |