Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106639.5 | NP_178108.1 | 1 - 2 | 577 | 79 | |
GenBank mRNA | AY065365.1 | - | 1 - 2 | 372 | 79 | |
AY096390.1 | - | 1 - 2 | 240 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |