Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106704.4 | NP_178171.1 | 2 - 3 | 411 | 85 | |
GenBank mRNA | Y10216.1 | - | 384 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |