Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334999.1 | NP_001319430.1 | 7 - 8 | 2550 | 64 | |
NM_001335000.1 | NP_001320420.1 | 8 - 9 | 2482 | 64 | ||
NM_001335001.1 | NP_001320419.1 | 8 - 9 | 2234 | 64 | ||
NM_106714.2 | NP_178181.1 | 8 - 9 | 2482 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |