Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335016.1 | NP_001321606.1 | 3 - 4 | 421 | 84 | |
NM_106731.3 | NP_178198.1 | 2 - 3 | 618 | 84 | ||
GenBank mRNA | AF165125.1 | - | 2 - 3 | 264 | 84 | |
BT029300.1 | - | 2 - 3 | 197 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |