Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111756.3 | NP_187533.1 | 1 - 2 | 275 | 69 | |
GenBank mRNA | AK117639.1 | - | 1 - 2 | 162 | 69 | |
BT004971.1 | - | 1 - 2 | 97 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |