Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB3R-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084658.2 | NP_001078127.1 | 3 - 4 | 443 | 68 | |
NM_001337814.1 | NP_001327447.1 | 2 - 3 | 751 | 68 | ||
NM_111771.4 | NP_566350.1 | 3 - 4 | 428 | 68 | ||
GenBank mRNA | AY142649.1 | - | 3 - 4 | 247 | 68 | |
AY519651.1 | - | 3 - 4 | 247 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |