Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337853.1 | NP_001326647.1 | 12 - 13 | 3033 | 73 | |
NM_111836.5 | NP_187612.5 | 11 - 12 | 2903 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |