Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035592.2 | NP_001030669.1 | 10 - 11 | 1198 | 81 | |
NM_001337876.1 | NP_001325521.1 | 10 - 11 | 1198 | 81 | ||
NM_001337877.1 | NP_001325520.1 | 11 - 12 | 1088 | 81 | ||
NM_111880.5 | NP_187656.1 | 11 - 12 | 1088 | 81 | ||
GenBank mRNA | AY088751.1 | - | 11 - 12 | 1083 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |