Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035592.2 | NP_001030669.1 | 7 - 8 | 941 | 143 | |
NM_001337876.1 | NP_001325521.1 | 7 - 8 | 941 | 143 | ||
NM_001337877.1 | NP_001325520.1 | 8 - 9 | 831 | 143 | ||
NM_111880.5 | NP_187656.1 | 8 - 9 | 831 | 143 | ||
GenBank mRNA | AY088751.1 | - | 8 - 9 | 826 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |