Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084749.1 | NP_001078218.1 | 5 - 6 | 580 | 85 | |
NM_113690.3 | NP_566824.1 | 7 - 8 | 865 | 85 | ||
GenBank mRNA | AY046004.1 | - | 7 - 8 | 796 | 85 | |
AY079315.1 | - | 7 - 8 | 722 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |