Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BR6OX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113917.4 | NP_566852.1 | 3 - 4 | 812 | 120 | |
GenBank mRNA | AB087801.1 | - | 3 - 4 | 710 | 120 | |
AY052655.1 | - | 3 - 4 | 762 | 120 | ||
AY063728.1 | - | 3 - 4 | 671 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |